Potassium cyanide 97+%

Supplier: Thermo Scientific Chemicals
Danger

L13273-22 L13273-36
AAL13273-22EA 73.3 USD
AAL13273-22 AAL13273-36
Potassium cyanide 97+%
Potassium cyanide

Purity: 97+% CAS No.: 151-50-8 Molecular Formula: CKN Formula Weight: 65.12 Melting Point: 634° Boiling Point: 1625° Merck Reference: 14,7626 UN No.: 1680 MDL No.: MFCD00011397

Formula: KCN
MW: 65.12 g/mol
Boiling Pt: 1625 °C
Melting Pt: 634 °C
Density: 1.52
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 14,7626

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR