Ethisterone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 17alpha-Ethynyltestosterone

J63580-06 J63580-14
AAJ63580-06EA 167 USD
AAJ63580-06 AAJ63580-14
Ethisterone 98%
Ethisterone

A progestogen that counteracts the estrogenic proliferative effect on the endometrium


Powder


Applications: A progestogen that counteracts the estrogenic proliferative effect on the endometrium

Formula: C₂₁H₂₈O₂
MW: 312.45 g/mol
Melting Pt: 263…269 °C
Storage Temperature: Ambient
MDL Number: MFCD00003656
CAS Number: 434-03-7
EINECS: 207-096-5
Merck Index: 14,3741

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR