Enalaprilat

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-1-((S)-2-(((S)-1-Carboxy-3-phenylpropyl)amino)propanoyl)pyrrolidine-2-carboxylic acid

J63392-MA J63392-MF
AAJ63392-MAEA 156 USD
AAJ63392-MA AAJ63392-MF
Enalaprilat
Enalaprilat

The active metabolite of enalapril


Crystalline solid


Applications: The active metabolite of enalapril

Formula: C₁₈H₂₈N₂O₇
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00865786
CAS Number: 76420-72-9
EINECS: 278-459-3
Merck Index: 14,3568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR