Sodium deoxycholate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate

J62288-14 J62288-22 J62288-30
AAJ62288-14EA 77.32 USD
AAJ62288-14 AAJ62288-22 AAJ62288-30
Sodium deoxycholate 97%
Sodium deoxycholate

Powder


Purity: 97% CAS No.: 302-95-4 Molecular Formula: C24H39NaO4 Formula Weight: 414.60 Physical Form: Powder Appearance: White Solubility: Soluble in water Beilstein Reference: 3581950 MDL No.: MFCD00064139

Formula: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Melting Pt: 357…365 °C
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4
EINECS: 206-132-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR