Harmaline ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61699-03
J61699-06
AAJ61699-03EA
51.4
USD
AAJ61699-03
AAJ61699-06
Harmaline ≥98%
Harmaline
CNS stimulant; may act through NMDA receptors. Reversible inhibitor of MAO-A
Crystals
Applications: CNS stimulant; may act through NMDA receptors. Reversible inhibitor of MAO-A
Formula:
C₁₃H₁₄N₂O Melting Pt: 239…241 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004955 CAS Number: 304-21-2 EINECS: 206-152-6 Merck Index: 14,4613 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...