Carboxy-PTIO potassium salt ≥98%

Supplier: Thermo Scientific Chemicals

J61316-MA J61316-MF
AAJ61316-MAEA 125 USD
AAJ61316-MA AAJ61316-MF
Carboxy-PTIO potassium salt ≥98%
Carboxy-PTIO potassium salt

Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase


Solid


Applications: Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase

Formula: C₁₄H₁₆KN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00216153
CAS Number: 148819-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR