1-(p-Toluenesulfonyl)imidazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(P-Toluenesulfonyl)imidazole, 1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole

H54191-06 H54191-14 H54191-22
AAH54191-06EA 43.5 USD
AAH54191-06 AAH54191-14 AAH54191-22
1-(p-Toluenesulfonyl)imidazole 98+%
1-(p-Toluenesulphonyl)imidazole

Purity: 98+% CAS No.: 2232-08-8 Molecular Formula: C10H10N2O2S Formula Weight: 222.26 Melting Point: 76-78° Beilstein Reference: 612451

Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00005285
CAS Number: 2232-08-8
EINECS: 218-771-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR