4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid
H53126-03
H53126-MD
AAH53126-03EA
170
USD
AAH53126-03
AAH53126-MD
4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid
Purity: 98%
CAS No.: 389621-84-5
Molecular Formula: C11H14BNO4
Formula Weight: 235.05
Melting Point: 126-132°
MDL No.: MFCD03411952
Formula:
C₁₁H₁₄BNO₄ Melting Pt: 126…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411952 CAS Number: 389621-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...