2-(4-Pyridyl)benzimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Pyridin-4-yl)-1H-benzo[d]imidazole

H51778-06 H51778-14
AAH51778-06EA 68.6 USD
AAH51778-06 AAH51778-14
2-(4-Pyridyl)benzimidazole 97%
2-(4-Pyridyl)benzimidazole

Purity: 97% CAS No.: 2208-59-5 Molecular Formula: C12H9N3 Formula Weight: 195.22 Odor: Odorless Melting Point: 216-221° MDL No.: MFCD00453948

Formula: C₁₂H₉N₃
Melting Pt: 216…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00453948
CAS Number: 2208-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR