4'-Bromo-2,2,2-trifluoroacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33627-03
H33627-06
AAH33627-03EA
135
USD
AAH33627-03
AAH33627-06
4'-Bromo-2,2,2-trifluoroacetophenone 97%
4'-Bromo-2,2,2-trifluoroacetophenone
Purity: 97%
CAS No.: 16184-89-7
Molecular Formula: C8H4BrF3O
Formula Weight: 253.02
Melting Point: 26-30°
Boiling Point: 95°/4mm
MDL No.: MFCD00191862
Formula:
C₈H₄BrF₃O Boiling Pt: 95 °C/4 mm Melting Pt: 26…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191862 CAS Number: 16184-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...