(E)-1-Ethoxyethene-2-boronic acid pinacolester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33367-03
H33367-06
AAH33367-03EA
168
USD
AAH33367-03
AAH33367-06
(E)-1-Ethoxyethene-2-boronic acid pinacolester 98%
(E)-1-Ethoxyethene-2-boronic acid pinacolester
Purity: 98%
CAS No.: 1201905-61-4
Molecular Formula: C10H19BO3
Formula Weight: 198.07
Refractive Index: 1.4460
Boiling Point: 50°/0.3mm
MDL No.: MFCD09702441
Formula:
C₁₀H₁₉BO₃ Boiling Pt: 50 °C/0.3 mm Storage Temperature: Ambient |
MDL Number:
MFCD09998813 CAS Number: 1201905-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...