Hydrogen chloride 1 M in ethyl acetate

Supplier: Thermo Scientific Chemicals
Danger

H32266-AE H32266-AP
AAH32266-AEEA 140.04 USD
AAH32266-AE AAH32266-AP
Hydrogen chloride 1 M in ethyl acetate
Hydrogen chloride

CAS No.: 7647-01-0 Molecular Formula: ClH Formula Weight: 36.46 Storage Temperature: 2°C to 8°C UN No.: 2924 MDL No.: MFCD00011324

Formula: HCl
MW: 36.46
Density: 0.900
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR