4-Cyano-3-fluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

H27710-14 H27710-03
AAH27710-06EA 246.28 USD
AAH27710-06 AAH27710-14 AAH27710-03
4-Cyano-3-fluorophenylboronic acid 97%
4-Cyano-3-fluorophenylboronic acid

Purity: 97% CAS No.: 843663-18-3 Molecular Formula: C7H5BFNO2 Formula Weight: 164.93 Melting Point: 300° MDL No.: MFCD03411549

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03411549
CAS Number: 843663-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR