Sodium methoxide in methanol ∼25% (w/w) in methanol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium methylate, Sodium methanolate

046585-0F 046585-AP
AA46585-0FEA 114.68 USD
AA46585-0F AA46585-AP
Sodium methoxide in methanol ∼25% (w/w) in methanol
Sodium methoxide

Liquid


CAS No.: 124-41-4 Molecular Formula: CH3ONa Formula Weight: 54.02 Flash Point: 29°(84°F) Physical Form: Liquid Density: 0.945 Appearance: Colorless to yellow Melting Point: -2° Boiling Point: 92° Beilstein Reference: 3592982 Merck Reference: 14,8643 UN No.: 1289 MDL No.: MFCD00012179

Formula: CH₃NaO
Boiling Pt: 92 °C
Melting Pt: –2 °C
Density: 0.945
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1289
ADR: 3,III
Merck Index: 14,8643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR