LB Broth, Ready-Made Powder, Thermo Scientific
Supplier: Thermo Fisher Scientific
J75852-A1 J75852-30
101174-120EA 181.94 USD
101174-120 101173-324
LB Broth, Ready-Made Powder, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
For the propagation of bacteria
Luria-Bertani Broth. Off-white free flowing powder.
pH(2.0%, H2O): 7.0 ± 0.2
Contents:
Casein Peptone: 10 gm/L
Yeast Extract: 5 gm/L
Sodium Chloride: 5 gm/L
To use, dissolve 20gm powder per liter of purified water. Autoclave.