Tetracycline hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T2525-25G
T2525-100G
TCT2525-25GEA
70.15
USD
TCT2525-25G
TCT2525-100G
Tetracycline hydrochloride ≥98.0% (by titrimetric analysis)
Tetracycline hydrochloride
CAS Number: 64-75-5
MDL Number: MFCD00078142
Molecular Formula: C22H24N2O8
Molecular Weight: 480.90
Purity/Analysis Method:
Formula:
C₂₂H₂₅ClN₂O₈ MW: 480.9 g/mol Melting Pt: 214 °C |
MDL Number:
MFCD00078142 CAS Number: 64-75-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...