Tetrabutylammonium fluoride 70-75% 70-75% in water

Supplier: TCI America
Danger

T1339-500G T1339-25G T1339-100G
TCT1339-500GEA 226 USD
TCT1339-500G TCT1339-25G TCT1339-100G
Tetrabutylammonium fluoride 70-75% 70-75% in water
Tetrabutylammonium fluoride

CAS Number: 429-41-4
MDL Number: MFCD00011747
Molecular Formula: C16H36FN
Molecular Weight: 261.47
Form: Clear Liquid
Color: Very Pale Yellow


Other Product Names / Synonyms: TBAF

Formula: C₁₆H₃₆FN
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
UN: 1760
ADR: 8,III
Merck Index: 14,09187
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e875015ff4b6695654a09d4cc49d582054c15cd&vid=93ee569b6d319050f7612c500003b35dace7c4fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Concentration (Nonaqueous Titration) 70.0 to 75.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR