1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose ≥98.0%
Supplier: TCI America
Synonyms:
1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose, (2S,3R,4R,5R)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate, β-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate, 2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate
CAS Number: 6974-32-9
MDL Number: MFCD00005357
Molecular Formula: C28H24O9
Molecular Weight: 504.49
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Glycosylation, Nucleic Acid Synthesis, Bioscience, Ribose, O-Substituted Sugars
Other Product Names / Synonyms: 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
Formula:
C₂₈H₂₄O₉ MW: 504.5 g/mol Melting Pt: 129 °C |
MDL Number:
MFCD00005357 CAS Number: 6974-32-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...