(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Naproxen, (S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid
CAS Number: 22204-53-1
MDL Number: MFCD00010500
Molecular Formula: C14H14O3
Molecular Weight: 230.26
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid, Naproxen
Formula:
C₁₄H₁₄O₃ MW: 230.26 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD00010500 CAS Number: 22204-53-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...