2-Methylcyclohexylamine (cis- and trans- mixture) ≥95.0% (by GC, titration analysis)

Supplier: TCI America

M0896-500ML M0896-25ML
TCM0896-500MLEA 305 USD
TCM0896-500ML TCM0896-25ML
2-Methylcyclohexylamine (cis- and trans- mixture) ≥95.0% (by GC, titration analysis)
2-Methylcyclohexylamine (cis- and trans- mixture)

CAS Number: 7003-32-9 MDL Number: MFCD00001492 Molecular Formula: C7H15N Molecular Weight: 113.20 Purity/Analysis Method: 95.0% (GC,T) Form: Clear Liquid Flash Point (°C): 22 Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1-Amino-2-methylcyclohexane

Formula: C₇H₁₅N
Density: 0.86 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00001492
CAS Number: 7003-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9acef8b5784a4096fe25fce605e265ca5984062&vid=a9acef8b5784a4096fe25fce605e265ca5984062&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR