DL-Malic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0020-25G M0020-500G
TCM0020-25GEA 31.63 USD
TCM0020-25G TCM0020-500G
DL-Malic acid ≥99.0% (by titrimetric analysis)

CAS Number: 6915-15-7 MDL Number: MFCD00064212 Molecular Formula: C4H6O5 Molecular Weight: 134.09 Purity/Analysis Method: 99.0% (T) Form: Crystal Boiling point (°C): 150 Melting point (°C): 130 Flash Point (°C): 203


Other Product Names / Synonyms: DL-Apple Acid

Formula: HOOCCH(OH)CH₂COOH
Boiling Pt: 150 °C
Melting Pt: 130 °C
Flash Pt: 203 °C
MDL Number: MFCD00064212
CAS Number: 617-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10f3b23d19b470de2b6eba07b2b382509a3100de&vid=10f3b23d19b470de2b6eba07b2b382509a3100de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR