Sodium 1-undecanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0349-25G I0349-5G
TCI0349-025GEA 229 USD
TCI0349-025G TCI0349-005G
Sodium 1-undecanesulfonate ≥98.0% (by titrimetric analysis)
Sodium 1-undecanesulphonate

[Reagent for Ion-Pair Chromatography] CAS Number: 5838-34-6 MDL Number: MFCD00053627 Molecular Formula: C11H24O3S Molecular Weight: 258.35 Purity/Analysis Method: 98.0% (T) Form: Crystal


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples Other Product Names / Synonyms: IPC-ALKS-11, 1-Undecanesulfonic Acid Sodium Salt

Formula: C₁₂H₂₅NaO₃S
MW: 258.36 g/mol
MDL Number: MFCD00053627
CAS Number: 5838-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d26cd2a4605d7accef8cd68136c4bf0af2f51eb&vid=78e4ba6e3f362327d5b32b0f2760a5ae2dca43a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR