Hydrochlorothiazide ≥97.0%

Supplier: TCI America

H1274-25G H1274-5G
TCH1274-25GEA 62 USD
TCH1274-25G TCH1274-5G
Hydrochlorothiazide ≥97.0%
Hydrochlorothiazide

CAS Number: 58-93-5 MDL Number: MFCD00051765 Molecular Formula: C7H8ClN3O4S2 Molecular Weight: 297.73 Purity/Analysis Method: 97.0% (HPLC,N) Form: Crystal Melting point (°C): 269 Lambda max.: 318 nm (H2O)


Product Categories: Bioscience, Other Pharmaceutical Ingredients

Formula: C₇H₈ClN₃O₄S₂
Melting Pt: 269 °C
MDL Number: MFCD00051765
CAS Number: 58-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcec211cb23722b8250cadf2a045da8da50cb030&vid=fcec211cb23722b8250cadf2a045da8da50cb030&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR