4-Hydrazinobenzenesulfonamide hydrochloride 98,0
Supplier: TCI America
Synonyms:
4-Hydrazinylbenzenesulphonamide hydrochloride, 4-Hydrazinylbenzenesulfonamide hydrochloride, 4-sulfonamidophenylhydrazine Hydrochloride, (4-sulphamoylphenyl)hydrazine HCl, 4-Sulphonamidophenylhydrazine Hydrochloride, (4-sulfamoylphenyl)hydrazine HCl
H1230-25G
H1230-5G
TCH1230-25GEA
99.4
USD
TCH1230-25G
TCH1230-5G
4-Hydrazinobenzenesulfonamide hydrochloride 98,0
4-Hydrazinobenzenesulfonamide hydrochloride
CAS Number: 17852-52-7
MDL Number: MFCD00052262
Molecular Formula: C6H9N3O2S
Molecular Weight: 223.68
Purity/Analysis Method:
Other Product Names / Synonyms: 4-Sulfonamidophenylhydrazine Hydrochloride
Formula:
C₆H₉N₃O₂S MW: 223.68 g/mol Melting Pt: 211 °C |
MDL Number:
MFCD00052262 CAS Number: 17852-52-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...