(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥97.0%
Supplier: TCI America
Synonyms:
H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl
H1172-1G
H1172-5G
TCH1172-1GEA
65.12
USD
TCH1172-1G
TCH1172-5G
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥97.0%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride
CAS Number: 1483-01-8
MDL Number: MFCD00012622
Molecular Formula: C7H16N4O2
Molecular Weight: 224.69
Purity/Analysis Method:
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-6-guanidinohexanoic Acid Hydrochloride, H-HomoArg-OH.HCl
Formula:
C₇H₁₆N₄O₂·HCl MW: 224.7 g/mol Melting Pt: 206 °C |
MDL Number:
MFCD00012622 CAS Number: 1483-01-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...