1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol ≥99.0%
Supplier: TCI America
Synonyms:
1,1,1,3,3,3-Hexafluoro-2-phenylpropan-2-ol, alpha,alpha-Bis(trifluoromethyl)benzyl Alcohol, 2,2,2,2',2',2'-Hexafluorocumyl Alcohol
H0724-25G
H0724-5G
TCH0724-025GEA
225.7
USD
TCH0724-025G
TCH0724-005G
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol ≥99.0%
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol
CAS Number: 718-64-9
MDL Number: MFCD00040983
Molecular Formula: C9H6F6O
Molecular Weight: 244.14
Purity/Analysis Method:
Other Product Names / Synonyms: 2,2,2,2',2',2'-Hexafluorocumyl Alcohol, alpha,alpha-Bis(trifluoromethyl)benzyl Alcohol
Formula:
C₉H₆F₆O MW: 244.14 g/mol Boiling Pt: 160 °C Density: 1.45 (20 °C) Flash Pt: 56 °C |
MDL Number:
MFCD00040983 CAS Number: 718-64-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...