Ethyl-3-methylvalerate ≥98.0%

Supplier: TCI America

E0803-5G E0803-25G
TCE0803-5GEA 48 USD
TCE0803-5G TCE0803-25G
Ethyl-3-methylvalerate ≥98.0%
Ethyl-3-methylvalerate

CAS Number: 5870-68-8 MDL Number: MFCD00041390 Molecular Formula: C8H16O2 Molecular Weight: 144.21 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 158 Flash Point (°C): 45 Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids Other Product Names / Synonyms: 3-Methylvaleric Acid Ethyl Ester, Ethyl 3-Methylpentanoate, 3-Methylpentanoic Acid Ethyl Ester

Formula: C₈H₁₆O₂
Boiling Pt: 158 °C
Density: 0.87 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00041390
CAS Number: 5870-68-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96733635cd9a6deba322e7b1ac9e8cf5ad93c81e&vid=96733635cd9a6deba322e7b1ac9e8cf5ad93c81e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR