DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon milt)

Supplier: TCI America

Synonyms: Thymonucleic acid sodium salt, DNA sodium salt

D3545-25G D3545-5G
TCD3545-25GEA 322.46 USD
TCD3545-25G TCD3545-5G
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon milt)
DNA sodium salt (Deoxyribonucleic acid sodium salt)

Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: Thymonucleic Acid Sodium Salt

MDL Number: MFCD00130921
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR