N,N-Dimethylaminomethylferrocene ≥97.0%

Supplier: TCI America

Synonyms: Ferrocenylmethyldimethylamine, (Dimethylaminomethyl)ferrocene

D1271-25G D1271-5G
TCD1271-025GEA 138 USD
TCD1271-025G TCD1271-005G
N,N-Dimethylaminomethylferrocene ≥97.0%
N,N-Dimethylaminomethylferrocene

CAS Number: 1271-86-9 MDL Number: MFCD00001433 Molecular Formula: C13H17FeN Molecular Weight: 243.13 Purity/Analysis Method: 97.0% (GC,T) Form: Liquid Flash Point (°C): 110 Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: Ferrocenylmethyldimethylamine

Formula: C₁₃H₁₇FeN
Density: 1.23 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001433
CAS Number: 1271-86-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d970a0d4b56a671b57cc4ac421f4c5c104171498&vid=d970a0d4b56a671b57cc4ac421f4c5c104171498&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR