p-Xylene-2-sulfonic acid hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0681-25G D0681-100G
TCD0681-25GEA 38 USD
TCD0681-25G TCD0681-100G
p-Xylene-2-sulfonic acid hydrate ≥98.0% (by HPLC, titration analysis)
p-Xylene-2-sulfonic acid hydrate

CAS Number: 609-54-1 MDL Number: MFCD00064858 Molecular Formula: C8H10O3S Molecular Weight: 186.23 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 86


Other Product Names / Synonyms: 2,5-Dimethylbenzenesulfonic Acid

Formula: C₈H₁₀O₃S
Melting Pt: 86 °C
MDL Number: MFCD00064858
CAS Number: 609-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4cb35c8bd546a9499b8c0a6c777746328a1a4eb&vid=b4cb35c8bd546a9499b8c0a6c777746328a1a4eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR