2-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2116-5G C2116-25G
TCC2116-5GEA 64.9 USD
TCC2116-5G TCC2116-25G
2-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
2-Chlorobenzhydrazide

CAS Number: 5814-05-1 MDL Number: MFCD00007597 Molecular Formula: C7H7ClN2O Molecular Weight: 170.60 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 118


Other Product Names / Synonyms: 2-Chlorobenzoylhydrazine, 2-Chlorobenzoic Hydrazide

Formula: C₇H₇ClN₂O
Melting Pt: 118 °C
MDL Number: MFCD00007597
CAS Number: 5814-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ceba6c18dc90b8fd6cff85b243c50b4cbf48b2e&vid=7ceba6c18dc90b8fd6cff85b243c50b4cbf48b2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR