Choline hydroxide 48 - 50% in water
Supplier: TCI America
CAS Number: 123-41-1
MDL Number: MFCD00002831
Molecular Formula: C5H15NO2
Molecular Weight: 121.18
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Hydroxyethyltrimethylammonium Hydroxide
Formula:
C₅H₁₅NO₂ Density: 1.08 (20 °C) |
MDL Number:
MFCD00002831 CAS Number: 123-41-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...