Value Plastics® Fitting, Polypropylene, Straight, Male Luer Lock to Plug Adapter; 1000/PK
Supplier: VALUE PLASTICS TE
VWR Item#MFLX30800-30BLK
Your Organization's Price:
Retrieving
Full material traceability to ensure process integrity.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Connection Type: Male luer lock to plug
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.