Value Plastics ABSML-6005-1 Fitting, White Nylon over Polypropylene, Male Luer with Rotating Lock Ring, 5/16" Hex to 1/4-28 UNF Thread, 1000/PK
Supplier: VALUE PLASTICS TE
VWR Item#MFLX15973-45BLK
Your Organization's Price:
Retrieving
Premium luer fittings for critical applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.