MasterSense® MP30 Pressure Sensor with ¹/₄" NPT Process Connection
Supplier: Avantor Fluid Handling
VWR Item#MFLX07526-80
Your Organization's Price:
Retrieving
High accuracy pressure transducers for plug and play connectivity with Masterflex® MasterSense® pump systems.
  • Add critical pressure monitoring to your workflow
  • All-stainless welded body and wetted parts
  • Rugged design for shock protection and excellent long-term stability
  • CE certificated
  • Sanitary models are designed for clean-in-place and sterilize-in-place
  • 0 to 30 psi
  • 4 to 20 mA output
  • CE Compliant
  • IP 65
MasterSense® MP30 pressure transducers utilize a thin-film-on-steel measurement principle to provide an accurate 0 to 30 psi output directly to MasterSense® pump systems. Sensors include a six foot cable with AUX plug adaptor for easy plug and play.
Designed for use with a ¹/₄" FNPT process tee connection (not included)
Install downstream of pump to detect pressure changes in your process. Perfect for tangential-Flow filtration, depth filtration, harvest, bulk transfer, chromatography and fill applications. Choose the model with the appropriate process connection for your application.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path. Model MFLX07526-81 ³/₄" sanitary connection not recommended for applications with high temperature variations.
<b>Certifications: </b>CE Certified.