CPC® Genderless Aseptic Fitting, 1/8" Barb, Polycarbonate, CNX17002HT; 25/PK
Supplier: Colder Products
VWR Item#MFLX20202-01
Your Organization's Price:
Retrieving
Provide easy, secure sterile connections even in nonsterile environments now in newly designed smaller sizes.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
  • Sterile micro-connectors eliminates the need to purchase, calibrate, validate, and maintain tube welding equiptment
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.<br /><br />These new MicroCNX connectors now reach smaller tubing sizes including 1/16" 1/8", and 3/32" IDs. <br /><br />Simply fit the connector half in your tubing line. The easy three-step PINCH-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. Pinch the protective cover off the connector. The audible “click” confirms a securely connected assembly. To complete the assembly, pull membrane strips directly away from the connector. <br /><br />MicroCNX fittings can be sterilized by gamma irradiation and autoclave. Each fitting meets USP Class VI and ADCF certifications.