Masterflex® Fitting, Polypropylene, Y-Adapter, Hose Barb Union, ¹/₂" Int.Ø; 25/Pack
Supplier: VWR International
VWR Item#MFLX40617-80
Your Organization's Price:
Retrieving
No tools needed for assembly.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: 3/4" ID ; 5/PK
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.<br />Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.