Masterflex® High-Efficiency Filter Bag for Heavy-Duty Housing, Polypropylene Absolute-Rated, Size 2, 1 µm
Supplier: VWR International
Wide range of options for temperature, flow rate, and filtration quality.
- Constructed of polypropylene (PP)
- FDA-compliant
Designed for 98% removal efficiency at the rated pore size, these bags offer three to four times the dirt-holding capacity of standard bags. In addition, they are FDA-compliant with respect to all regulations governing food-grade materials. These polypropylene absolute-rated filter bags are rated for a temperature of 200 °F (93 °C) maximum.