Masterflex® Transfer Tubing, Premo Flex®, 1/2" ID x 0.85" OD; 100 Ft
Supplier: VWR International
VWR Item#MFLX95953-03
Your Organization's Price:
Retrieving
Resists weather and ozone.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Outstanding resistance to heat and ozone
  • Maximum pressure: 250 psi (17.2 bar)
  • Minimum bend radius: 4"
  • Inner diameter: 1/2"
Masterflex® Premp Flex® transfer tubing is ideal for use with fuels such as petroleum oils, gasoline, kerosene, and lubricating oils, while maintaining tube integrity. The Buna-N tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube meets RMA Class A high oil resistance and nonconductive at 1000 VDC certification. The tube cover meets Class B medium oil resistance and nonconductive at 1000 VDC certification.