Masterflex® Fitting, Nylon, Straight, Hose Barb Reducer, 1/4" ID x 3/16" ID; 100/PK
Supplier: VWR International
VWR Item#MFLX30626-30
Your Organization's Price:
Retrieving
Best for low-pressure applications.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperature range −50 to +250 °F (−46 to +121 °C)
  • Connection Type: 1/4" ID x 3/16" ID