LeviFlow LFSC-12D-007 Clamp-On Ultrasonic Flow Sensor, 20 L/min
Supplier: LeviFlow
No moving parts—no particle generation—no contamination.
- Starter kit and flow sensor required to build a complete system
- Use on your existing silicone or C-Flex tubing
- Obtain flow measurement without intrusion
- Reusable flow sensors—choose from 4-, 20-, or 80-LPM models
These flow systems are ideal for sterile applications. Flow sensors fit over your existing silicone or C-Flex tubing lines and are easily reusable—remove sensor clamp, dispose of flexible tubing, and fix sensor over a new line.<br />These non-invasive flow measurement systems have no moving parts to contaminate or generate particulates, and are accurate to 3%.
<b>Delivery information: </b>Includes water tight connection box.