Masterflex® Sanitary Tubing Adapter, PVDF; 3/4" Mini to Female Luer Lock
Supplier: VWR International
VWR Item#MFLX40619-96
Your Organization's Price:
Retrieving
Attach with just a twist!
  • Choose between polypropylene (PP) and PVDF material options
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.