Masterflex® Fitting, PVDF, Straight, Hose Barb Unions, 5/8" ID; 10/PK
Supplier: VWR International
Quick and easy assembly without tools.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Single-use custom lengths and configurations available to minimize downtime
- Ultra smooth inner surface results in fewer extractables
- Connection Type: 1/2" ID