Masterflex® L/S® Precision Pump Tubing, BioPharm Platinum-Cured Silicone, L/S 25; 25 ft
Supplier: VWR International
VWR Item#MFLX96420-25
Your Organization's Price:
Retrieving
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 25
<B>Applications</B>: Pharmaceutical, cosmetic, research, and auto-analysis applications<br /><B>Characteristics</B>: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent