Masterflex® L/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 17; 12"/Pk
Supplier: VWR International
VWR Item#MFLX96242-17
Your Organization's Price:
Retrieving
High-performance pump tubing for challenging applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Custom extruded to fit Masterflex® pumps
  • Lot-to-lot consistency provides superior accuracy and repeatability
  • Tubing Size: L/S 17
<B>Applications</B>: Chemical and pharmaceutical applications<br /><B>Characteristics</B>: Enhanced chemical resistance, including aggressive organic solvents, spallation-free, low gas permeability, opaque white<br />Gore® STA-PURE® PFL-Series Pump Tubing is your ideal choice for high purity or long-running, critical pharmaceutical and biopharmaceutical processing applications. The patented composite (perfluoroelastomer) and ePTFE (polytetrafluoroethylene), results in a tube that offers high flex durability for greater resistance to rupture—even at applications up to 60 psig. Long-term pump testing proves that Gore® STA-PURE® PFL-Series Pump Tubing lasts hundreds of hours with consistent and repeatable flow rates resulting in one saving time and reducing maintenance by having fewer tubing changes.<br /><br />If one is looking for a flow stable / process stable tubing that withstands CIP (clean-in-place) or SIP (steam-in-place) cleaning—look no further than this tubing! <br /><br />Gore® STA-PURE® Pump tubing - Series PFL is available as single tube elements. Recommended for use in L/S® and I/P® standard and Easy-Load® pump heads, and L/S® Easy-Load® II pump heads.<br /><B>NOTE: </B> There is an engineering questionnaire vetting application specific details before the order can be processed.