Dow Corning Transfter Tubing, Pharm-80 Platinum-Cured Silicone, 1/2" ID×3/4" OD; 50 Ft
Supplier: Liveo
VWR Item#MFLX96109-11
Your Organization's Price:
Retrieving
Pharma tubing remains flexible and resilient for long service life.
<B>Applications:</B> Pharmaceutical, biotechnology, and ultra-pure fluid processing.<br /><B>Characteristics:</B> Semi-transparent platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex or other organic additives. Pharma tubing is hydrophobic, and remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicone to silicone tubing. Durometer, shore A rating of 75.<br /><B>Certification:</B> Elastomer exceeds test guidelines for USP Class VI Plastics; tubing itself is FDA compliant, and meets 3A Sanitary Standards and European Pharmacopoeia (EP) Requirements. Pharma tubing is produced under Bulk Pharmaceutical (BP) current GMP principals.