Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/4" ID×7/16" OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX95702-14
Your Organization's Price:
Retrieving
Reduced particle entrapment with ultra-smooth inner surface.
  • Minimal extractables help maintain fluid integrity
Formulation: TYGON® 3350 silicone, platinum cured.<br />Applications: Pharmaceutical, cosmetic, industrial, and auto-analysis applications.<br />Characteristics: Ultra-smooth inner surface reduces particle entrapment and minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; is odorless, tasteless, and non-toxic. Transparent.<br />Certification: Elastomer is FDA-compliant (21 CFR 177.2600); meets USP Class VI requirements, NSF criteria and EP 3.2.9 requirements. Tubing is manufactured according to GMP.<br />Temperature range: –112 to 392 °F (–80 to 200 °C)<br />Sterilize: With ethylene oxide, gamma irradiation, or autoclave for 30 min, 15 psi.