Masterflex® Ismatec® Pump Tubing, 2-Stop, Solva™, 2.06 mm ID; 12/PK
Supplier: VWR International
VWR Item#MFLX95606-42
Your Organization's Price:
Retrieving
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Ideal for chromatography applications and for the transfer of fuels, distillates, lubricants, and glycol-based coolants
  • Resistant to ozone and UV light; resistant to cracking, swelling, and hardening
  • <B>Appearance:</B> Yellow, translucent
  • <B>Temperature:</B> –30 to 85 °C
  • Inner diameter: 2.06 mm
The overall length of Solva 95606-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long. Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.