Masterflex® Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, 3/4" Mini x 1/2" NPT(F)
Supplier: VWR International
VWR Item#MFLX31200-13
Your Organization's Price:
Retrieving
Maintain sanitary conditions.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® connections
  • Excellent chemical resistance; low leachable characteristics
  • Connection Type: 3/4" mini to 1/2" NPT(F)
These fittings are ideal for use with the Masterflex L/S tubing pumps.