Masterflex® Transfer Tubing, Peroxide-Cured Silicone, 1/32" ID x 3/32" OD; 25 Ft
Supplier: VWR International
VWR Item#MFLX06411-60
Your Organization's Price:
Retrieving
No need to worry about the heat - use for applications up to 460 °F.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 1/32" ID x 3/32" OD
Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications<br />Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent<br />Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP.<br />Temperature range: -60 to 460 °F (-51 to 238 °C)<br />Sterilize: Autoclave or gamma irradiation